Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a23/3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013105.2 | NP_037237.2 | 516 | |||
GenBank mRNA | BC088163.1 | - | 516 | 7370410 | ||
D13912.1 | - | 5, 6 | 511 | |||
D29967.1 | - | 513 | ||||
FQ211106.1 | - | 526 | ||||
FQ218135.1 | - | 1248 | ||||
L24207.1 | - | 503 | ||||
X96721.1 | - | 513 |
Gene Symbol: LOC100910877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751127.3 | XP_003751175.1 | 5, 6 | 541 |