Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013108.2 | NP_037240.2 | 1, 2 | 1391 | ||
XM_008771315.1 | XP_008769537.1 | 1, 2 | 1716 | |||
GenBank mRNA | M74716.1 | - | 1, 2 | 1390 | ||
S56481.1 | - | 1, 2 | 1182 | |||
S73473.1 | - | 1, 2 | 1221 |
Set Membership: |
![]() |