Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a23/3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013105.2 | NP_037237.2 | 569 | |||
GenBank mRNA | BC088163.1 | - | 569 | 7370410 | ||
D13912.1 | - | 6 | 564 | |||
D29967.1 | - | 566 | ||||
FQ211106.1 | - | 579 | ||||
FQ218135.1 | - | 1301 | ||||
L24207.1 | - | 556 | ||||
X96721.1 | - | 566 |
Gene Symbol: LOC100910877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751127.3 | XP_003751175.1 | 6 | 594 |