Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013110.2 | NP_037242.2 | 3, 4 | 244 | ||
XM_006232139.2 | XP_006232201.1 | 3, 4 | 3090 | |||
XM_008760860.1 | XP_008759082.1 | 3, 4 | 3090 | |||
GenBank mRNA | AF010464.1 | - | 3, 4 | 253 | ||
AF367210.1 | - | 3, 4 | 244 | |||
FQ226549.1 | - | 814 |