Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013131.1 | NP_037263.1 | 3159 | |||
GenBank mRNA | M36074.1 | - | 3159 |