Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013187.1 | NP_037319.1 | 3 | 483 | ||
GenBank mRNA | J03806.1 | - | 3 | 483 |