Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016989.2 | NP_058685.1 | 5 | 783 | ||
XM_006245672.2 | XP_006245734.1 | 6 | 1479 | |||
XM_006245673.2 | XP_006245735.1 | 6 | 1855 | |||
XM_006245674.2 | XP_006245736.1 | 6 | 1808 | |||
XM_008767418.1 | XP_008765640.1 | 6 | 1432 | |||
XM_008767419.1 | XP_008765641.1 | 4 | 829 |