Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017012.1 | NP_058708.1 | 2 | 372 | ||
GenBank mRNA | D10891.1 | - | 2 | 372 | ||
EU559292.1 | - | 1 | 105 |
Set Membership: |
![]() |