Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013215.1 | NP_037347.1 | 5 | 702 | ||
GenBank mRNA | AF045464.1 | - | 5 | 702 | ||
BC078872.1 | - | 5 | 696 | 7127297 | ||
BC089811.1 | - | 5 | 665 | 7126881 | ||
X74673.1 | - | 5 | 699 |