Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013220.1 | NP_037352.1 | 2, 3 | 269 | ||
XM_008760363.1 | XP_008758585.1 | 3, 4 | 329 | |||
GenBank mRNA | BC072699.1 | - | 2, 3 | 261 | 7105899 | |
L81174.1 | - | 2, 3 | 219 | |||
U50736.1 | - | 2, 3 | 269 |