Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013200.1 | NP_037332.1 | 11, 12 | 1399 | ||
XM_006242180.2 | XP_006242242.1 | 11, 12 | 1855 | |||
GenBank mRNA | BC085761.1 | - | 11, 12 | 1411 | 7193798 | |
D43623.1 | - | 10, 11 | 1386 |