Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017031.2 | NP_058727.2 | 1 | 628 | ||
GenBank mRNA | U95748.1 | - | 1 | 628 |