Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113328.1 | NP_001106799.1 | 7 | 977 | ||
NM_001113329.1 | NP_001106800.1 | 8 | 892 | |||
NM_001113332.1 | NP_001106803.1 | 7 | 829 | |||
NM_001113334.1 | NP_001106805.1 | 7 | 666 | |||
NM_017032.1 | NP_058728.1 | 7 | 584 | |||
XM_008760721.1 | XP_008758943.1 | 7 | 1985 | |||
XM_008760722.1 | XP_008758944.1 | 2 | 7688 | |||
XM_008760723.1 | XP_008758945.1 | 3 | 328 | |||
GenBank mRNA | AF031373.1 | - | 7 | 977 | ||
AF536974.1 | - | 2 | 300 | |||
AF536979.1 | - | 8 | 892 | |||
AY388961.1 | - | 7 | 602 | |||
EF102484.1 | - | 7 | 829 | |||
EF121818.1 | - | 7 | 666 | |||
L27059.1 | - | 4 | 303 | |||
L27060.1 | - | 2 | 221 | |||
U09455.1 | - | 2 | 470 | |||
U09456.1 | - | 3 | 328 | |||
U09457.1 | - | 7 | 584 |