Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017080.2 | NP_058776.2 | 6 | 1100 | ||
XM_006250471.2 | XP_006250533.1 | 7 | 881 | |||
XM_006250473.2 | XP_006250535.1 | 7 | 916 | |||
GenBank mRNA | AF542063.1 | - | 5 | 700 | ||
AY387051.1 | - | 371 | ||||
BC078865.1 | - | 6 | 921 | 7109256 | ||
FQ209646.1 | - | 6 | 867 | |||
FQ209829.1 | - | 6 | 867 | |||
FQ210302.1 | - | 6 | 865 | |||
FQ210563.1 | - | 6 | 865 | |||
FQ210596.1 | - | 6 | 867 | |||
FQ211059.1 | - | 6 | 864 | |||
FQ211404.1 | - | 6 | 867 | |||
FQ212763.1 | - | 6 | 868 | |||
FQ214152.1 | - | 6 | 867 | |||
FQ214397.1 | - | 7 | 923 | |||
FQ218558.1 | - | 6 | 865 | |||
FQ218584.1 | - | 6 | 867 | |||
FQ219123.1 | - | 6 | 867 | |||
FQ219241.1 | - | 6 | 866 | |||
FQ219385.1 | - | 6 | 865 | |||
FQ219394.1 | - | 6 | 866 | |||
FQ219517.1 | - | 6 | 867 | |||
FQ219567.1 | - | 6 | 876 | |||
J05107.1 | - | 6 | 845 |