Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320972.1 | NP_001307901.1 | 3 | 897 | ||
NM_001320973.1 | NP_001307902.1 | 3 | 897 | |||
NM_003666.3 | NP_003657.1 | 3 | 897 | |||
XM_005245561.4 | XP_005245618.1 | 3 | 804 | |||
XM_011510092.2 | XP_011508394.1 | 3 | 800 | |||
XM_017002645.1 | XP_016858134.1 | 3 | 767 | |||
GenBank mRNA | AF288161.1 | - | 3 | 787 | ||
AK223533.1 | - | 3 | 759 | |||
AK291725.1 | - | 3 | 564 | |||
BC020716.1 | - | 3 | 546 | 4723421 | ||
U79751.1 | - | 3 | 786 |