Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017232.3 | NP_058928.3 | 3 | 254 | ||
GenBank mRNA | AF233596.1 | - | 378 | |||
AY157736.1 | - | 3 | 254 | |||
L20085.1 | - | 3 | 254 | |||
L25925.1 | - | 3 | 260 | |||
S67722.1 | - | 378 | ||||
U03389.1 | - | 378 | ||||
U04300.1 | - | 3 | 254 |