Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167578.1 | NP_001161050.1 | 9, 10 | 1290 | ||
GenBank mRNA | AK076614.1 | - | 9, 10 | 1236 | ||
BC061173.1 | - | 9, 10 | 1261 | 6772123 |