Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 9 | 4087 | ||
NM_147187.2 | NP_671716.2 | 10 | 4000 | |||
NR_027140.1 | 9 | 3894 | ||||
XM_017013944.1 | XP_016869433.1 | 9 | 3875 | |||
GenBank mRNA | AF016266.1 | - | 9 | 3910 | ||
BX537617.1 | - | 4 | 3192 | |||
BX538104.1 | - | 9 | 3917 | |||
BX538179.1 | - | 4 | 3192 |