Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195531.1 | NP_001182460.1 | 1, 2 | 179 | ||
GenBank mRNA | BC147108.1 | - | 1, 2 | 179 | 9056151 | |
BC147109.1 | - | 1, 2 | 179 | 9056141 |