Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033205.3 | NP_001028377.1 | 2 | 260 | ||
GenBank mRNA | AK138180.1 | - | 2 | 215 | ||
AK159025.1 | - | 2 | 261 | |||
BC150867.1 | - | 2 | 226 | 9087778 | ||
BC150873.1 | - | 2 | 226 | 9087784 |