Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017212.2 | NP_058908.2 | 9, 10 | 1050 | ||
XM_008768268.1 | XP_008766490.1 | 13, 14 | 2223 | |||
XM_008768269.1 | XP_008766491.1 | 12, 13 | 2136 | |||
XM_008768270.1 | XP_008766492.1 | 12, 13 | 2131 | |||
XM_008768271.1 | XP_008766493.1 | 11, 12 | 2049 | |||
XM_008768272.1 | XP_008766494.1 | 12, 13 | 2024 | |||
XM_008768273.1 | XP_008766495.1 | 11, 12 | 1931 | |||
XM_008768274.1 | XP_008766496.1 | 12, 13 | 1457 | |||
XM_008768275.1 | XP_008766497.1 | 11, 12 | 1370 | |||
XM_008768277.1 | XP_008766499.1 | 11, 12 | 1256 | |||
XM_008768278.1 | XP_008766500.1 | 10, 11 | 1169 | |||
XM_008768279.1 | XP_008766501.1 | 10, 11 | 1164 | |||
XM_008768280.1 | XP_008766502.1 | 9, 10 | 1076 | |||
XM_008768281.1 | XP_008766503.1 | 8, 9 | 989 | |||
XM_008768282.1 | XP_008766504.1 | 14, 15 | 2278 | |||
GenBank mRNA | BC126095.1 | - | 8, 9 | 957 | 8367931 | |
M84156.1 | - | 12, 13 | 1881 | |||
X79321.1 | - | 9, 10 | 1137 |