Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Synpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177340.2 | NP_796314.2 | 3 | 4460 | ||
GenBank mRNA | AK034012.1 | - | 2 | 1991 | ||
AK049810.1 | - | 2 | 2052 | |||
AK154958.1 | - | 3 | 4455 | |||
AK172163.1 | - | 3 | 4411 | |||
BC065127.1 | - | 3 | 4606 | |||
BC100364.1 | - | 1 | 1915 |