Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033219.3 | NP_001028391.2 | 8 | 4138 | ||
NM_001168255.1 | NP_001161727.1 | 7 | 3216 | |||
GenBank mRNA | AK034301.1 | - | 1 | 678 | ||
AK170738.1 | - | 7 | 3221 | |||
AK173104.1 | - | 4 | 2395 | |||
BC056501.1 | - | 8 | 3735 | 5720940 | ||
BC058722.1 | - | 7 | 3134 |