Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rimklb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027664.1 | NP_081940.1 | 6 | 3783 | ||
GenBank mRNA | AK173135.1 | - | 7 | 3790 |