Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033227.2 | NP_001028399.1 | 12 | 1418 | ||
GenBank mRNA | AK082798.1 | - | 1 | 74 | ||
AK144033.1 | - | 12 | 1419 | |||
AK165337.1 | - | 11 | 1214 | |||
BC141325.1 | - | 11 | 1211 | 9055937 |