Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009592.1 | NP_033722.1 | 16 | 3326 | ||
GenBank mRNA | BC035534.1 | - | 13 | 2954 | ||
BC151037.1 | - | 16 | 3326 | 9087950 |