Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010246.1 | NP_034376.1 | 1 | 1375 | ||
GenBank mRNA | AF033585.2 | - | 1 | 1230 | ||
AK021164.1 | - | 1 | 1559 | |||
Y17709.1 | - | 1 | 928 |
Set Membership: |
![]() |