Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010397.4 | NP_034527.1 | 1 | 147 | ||
GenBank mRNA | AK161744.1 | - | 1 | 146 | ||
M35243.1 | - | 1 | 116 | |||
M35245.1 | - | 1 | 147 | |||
M35247.1 | - | 1 | 128 |
Gene Symbol: H2-T10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161731.1 | - | 1 | 158 | ||
BC021751.1 | - | 142 | 4165944 | |||
BC021760.1 | - | 166 | 5055493 |