Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Htr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017254.1 | NP_058950.1 | 1 | 144 | ||
GenBank mRNA | M30705.1 | - | 1 | 144 | ||
M64867.1 | - | 550 | ||||
X13971.1 | - | 686 |
Set Membership: |
G-protein coupled receptor |