Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pea15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011063.2 | NP_035193.1 | 4 | 1521 | ||
GenBank mRNA | AK077421.1 | - | 4 | 1460 | ||
AK089070.1 | - | 4 | 1449 | |||
AK153141.1 | - | 4 | 1454 | |||
AK153493.1 | - | 4 | 1452 | |||
AK161635.1 | - | 4 | 1436 | |||
AK161977.1 | - | 4 | 1450 | |||
BC038282.1 | - | 4 | 1429 | 4500957 | ||
BC092079.1 | - | 2 | 633 | |||
G49234.1 | - | 4 | 1443 | |||
L31958.1 | - | 1 | 757 |