Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020255.3 | NP_064651.2 | 2 | 489 | ||
GenBank mRNA | AF106473.1 | - | 1 | 312 | ||
AF220501.1 | - | 2 | 316 | |||
AK009115.1 | - | 2 | 352 | |||
AK088253.1 | - | 2 | 319 | |||
AK220179.1 | - | 1 | 348 | |||
BC010724.1 | - | 1 | 296 | 2812488 | ||
CA451199.1 | - | 2 | 318 |