Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177667.1 | NP_001171138.1 | 16 | 2275 | ||
NM_001177668.1 | NP_001171139.1 | 16 | 2275 | |||
GenBank mRNA | AK132224.1 | - | 16 | 2159 | ||
AK141149.1 | - | 16 | 2156 | |||
AK147317.1 | - | 16 | 2159 | |||
BC150941.1 | - | 16 | 2111 | 9087852 | ||
BC158015.1 | - | 16 | 2111 | 9007321 | ||
BC172095.1 | - | 16 | 2111 | 9054381 |