Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017281.1 | NP_058977.1 | 4 | 198 | ||
GenBank mRNA | BC063170.1 | - | 4 | 218 | 6921964 | |
X53304.1 | - | 4 | 198 | |||
X55986.1 | - | 4 | 154 |