Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163103.1 | NP_001156575.1 | 3, 4 | 278 | ||
GenBank mRNA | AK133227.1 | - | 3, 4 | 257 | ||
CK137158.1 | - | 3, 4 | 278 |