Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111045.1 | NP_001104515.1 | 8 | 1592 | ||
NM_001111046.1 | NP_001104516.1 | 8 | 1474 | |||
NM_001111047.1 | NP_001104517.1 | 8 | 1471 | |||
NM_003914.3 | NP_003905.1 | 8 | 1595 | |||
XM_011535294.2 | XP_011533596.1 | 8 | 2426 | |||
XM_011535295.2 | XP_011533597.1 | 8 | 2423 | |||
XM_011535296.2 | XP_011533598.1 | 8 | 1251 | |||
GenBank mRNA | AK225146.1 | - | 6 | 940 | ||
AK301897.1 | - | 8 | 1367 | |||
AK314261.1 | - | 8 | 1595 | |||
AK316392.1 | - | 8 | 1394 | |||
BC035479.1 | - | 8 | 1341 | |||
BC036346.1 | - | 8 | 1341 | 5172478 | ||
BT019577.1 | - | 8 | 1236 | |||
U66838.1 | - | 8 | 1374 | |||
U97680.1 | - | 7 | 1084 |