Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102430.1 | NP_001095900.1 | 25 | 3828 | ||
GenBank mRNA | AK134369.1 | - | 1 | 102 |