Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033249.3 | NP_001028421.1 | 5 | 847 | ||
GenBank mRNA | AK038614.1 | - | 5 | 628 | ||
AK133380.1 | - | 5 | 849 | |||
BC056221.1 | - | 3 | 436 |