Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 8, 9 | 3072 | ||
GenBank mRNA | AK045780.1 | - | 6, 7 | 1041 | ||
AK122230.1 | - | 4, 5 | 523 | |||
BC099524.1 | - | 2, 3 | 155 | |||
BC137913.1 | - | 8, 9 | 3078 | 8860932 |