Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 3 | 1726 | ||
GenBank mRNA | AK135686.1 | - | 1 | 478 | ||
BC011226.1 | - | 3 | 1721 | |||
BC019710.1 | - | 1 | 206 | |||
BC032205.1 | - | 3 | 1740 | |||
BC038053.1 | - | 1 | 136 | |||
BC137913.1 | - | 3 | 1732 | 8860932 |