Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 15 | 7536 | ||
GenBank mRNA | AK010013.1 | - | 1 | 1769 | ||
AK010180.1 | - | 1 | 1710 | |||
AK017402.1 | - | 1 | 1290 |