Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081139.1 | NP_001074608.1 | 5 | 642 | ||
GenBank mRNA | AK028691.1 | - | 5 | 432 | ||
AK039772.1 | - | 5 | 397 | |||
AK046624.1 | - | 5 | 451 | |||
BC138276.1 | - | 5 | 444 | 8861295 | ||
BC145228.1 | - | 5 | 448 | 9053780 | ||
BC145750.1 | - | 5 | 444 | 40131053 | ||
BC157973.1 | - | 5 | 444 | 9007295 |