Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033254.3 | NP_001028426.2 | 10 | 2416 | ||
NM_001145854.1 | NP_001139326.1 | 9 | 1941 | |||
GenBank mRNA | AK028788.1 | - | 7 | 2312 | ||
AK145601.1 | - | 9 | 1915 | |||
AK158913.1 | - | 10 | 2690 | |||
AK158944.1 | - | 10 | 2690 | |||
BC059912.1 | - | 10 | 2222 | |||
BC079572.1 | - | 10 | 2413 | |||
BC150754.1 | - | 10 | 2306 | 9087665 |