Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033261.2 | NP_001028433.2 | 35 | 6278 | ||
GenBank mRNA | AK133182.1 | - | 7 | 869 | ||
AK161448.1 | - | 7 | 847 | |||
AK172969.1 | - | 33 | 4978 | |||
BC033596.1 | - | 13 | 1608 | |||
BC046488.1 | - | 21 | 3123 | |||
BC061510.1 | - | 23 | 3531 | |||
BC139124.1 | - | 35 | 6135 | 8862146 |