Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033261.2 | NP_001028433.2 | 15 | 3451 | ||
GenBank mRNA | AK145652.1 | - | 15 | 3454 | ||
AK172969.1 | - | 13 | 2151 | |||
BC046488.1 | - | 1 | 296 | |||
BC061510.1 | - | 3 | 705 | |||
BC139124.1 | - | 15 | 3308 | 8862146 |