Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017325.1 | NP_059021.1 | 2, 3 | 835 | ||
XM_008768598.1 | XP_008766820.1 | 2, 3 | 2637 | |||
XM_008768599.1 | XP_008766821.1 | 2, 3 | 2637 | |||
GenBank mRNA | L35271.1 | - | 2, 3 | 835 |