Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033272.1 | NP_001028444.1 | 2 | 565 | ||
GenBank mRNA | BC138455.1 | - | 2 | 522 | 8861475 | |
BC145285.1 | - | 2 | 522 | 9053839 | ||
CD352004.1 | - | 2 | 540 | |||
CF182771.1 | - | 1 | 413 |