Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017236.1 | NP_058932.1 | 3 | 340 | ||
GenBank mRNA | BC063171.1 | - | 3 | 357 | 6888051 | |
FQ211637.1 | - | 3 | 301 | |||
FQ212424.1 | - | 3 | 323 | |||
FQ215504.1 | - | 3 | 351 | |||
FQ224857.1 | - | 3 | 351 | |||
X71873.1 | - | 3 | 315 | |||
X75253.1 | - | 3 | 340 |