Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162883.1 | NP_001156355.1 | 3 | 736 | ||
NM_173786.1 | NP_776147.1 | 3 | 788 | |||
GenBank mRNA | BC020489.1 | - | 3 | 766 | 2649139 | |
BC057684.1 | - | 3 | 796 | 3675306 |