Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178923.3 | NP_849254.2 | 19 | 2896 | ||
GenBank mRNA | AK045266.1 | - | 19 | 2884 | ||
BC023734.1 | - | 16 | 2297 | |||
BC024860.1 | - | 1 | 24 | |||
BC041782.1 | - | 16 | 2268 | |||
BC057592.1 | - | 19 | 2896 | 6417362 |