Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Znhit6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081094.1 | NP_001074563.1 | 8 | 1318 | ||
GenBank mRNA | AK010555.1 | - | 7 | 663 | ||
AK148729.1 | - | 8 | 1295 | |||
AK167835.1 | - | 9 | 741 | |||
AK168844.1 | - | 9 | 734 | |||
BC147498.1 | - | 8 | 1204 | 9056360 | ||
BC150966.1 | - | 8 | 1204 | 9087878 | ||
BC150967.1 | - | 8 | 1204 | 9087879 | ||
BC151198.1 | - | 8 | 1204 | 9088086 |